missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse bFGF / FGF2 Protein
A DNA sequence encoding the Mus musculus (Mouse) bFGF / FGF2, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP8538-ec-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_032032.1 | |
bFGF / FGF2 Protein | |
Research Use Only | |
FGF2 | |
Recombinant Protein | |
E. coli | |
PALPEDGGAA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHV KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTEE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS | |
Greater than 90% as determined by SDS-PAGE. |
14173 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |