missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Leptin Protein

A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10756-5mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15995068

  • 665.00€ / 5mg
Expédition estimée: 28-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Leptin Protein
Research Use Only
Pegylated mouse Leptin is capable of stimulatng proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Pegylated mouse Leptin in vitro activity is only slightly lower than the non-pegylated antagonist but in vivo it has profound weight reducing effect (as compared to the non-pegylated leptin), resulting mainly from reduced food intake.
Mouse
Untagged
The mouse Leptin was lyophilized from a concentrated (0.65 mg/ml) solution containing 0.003mM NaHCO3.
5 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MCWRPLCRFL WLWSYLSYVQ AVPIQKVQDD TKTLIKTIVT RINDISHTQS VSAKQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQVLTSLP SQNVLQIAND LENLRDLLHL LAFSKSCSLP QTSGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDVSPEC
Greater than 99.0% as determined by:(a) Analysis by Gel-Filtration. (b) Analysis by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions