missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse RELM g Protein
A cDNA sequence encoding the RELM g was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13295-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
245195 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MEGTLESIVE KKVKELLANR DDCPSTVTKT FSCTSITASG RLASCPSGMT VTGCACGYGC GSWDIRDGNT CHCQCSTMDW ATARCCQLA | |
Greater than 95.0% as determined by SDS-PAGE. |
RELM g Protein | |
Research Use Only | |
Retnlg | |
Mouse | |
Untagged | |
Lyophilized from a sterile filtered solution containing 0.1 % trifluoroacetic acid (TFA). |