Learn More
enQuireBio™ Recombinant Mouse Roundabout homolog 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Roundabout homolog 1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP10099-ma-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_062286.2 | |
Roundabout homolog 1 Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
19876 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Robo1 | |
Recombinant Protein | |
Mammal | |
YRHRKKRNGL TSTYAGIRKV PSFTFTPTVT YQRGGEAVSS GGRPGLLNIS EPATQPWLAD TWPNTGNNHN DCSINCCTAG NGNSDSNLTT YSRPADCIAN YNNQLDNKQT NLMLPESTVY GDVDLSNKIN EMKTFNSPNL KDGRFVNPSG QPTPYATTQL IQANLSNNMN NGAGDSSEKH WKPPGQQKPE VAPIQYNIME QNKLNKDYRA NDTIPPTIPY NQSYDQNTGG SYNSSDRGSS TSGSQGHKKG ARTPKAPKQG GMNWADLLPP PPAHPPPHSN SEEYNMSVDE SYDQEMPCPV PPAPMYLQQD ELQEEEDERG PTPPVRGAAS SPAAVSYSHQ STATLTPSPQ EELQPMLQDC PEDLGHMPHP PDRRRQPVSP PPPPRPISPP HTYGYISGPL VSDMDTDAPE EEEDEADMEV AKMQTRRLLL RGLEQTPASS VGDLESSVTG SMINGWGSAS EEDNISSGRS SVSSSDGSFF TDADFAMS-Q AVAAAAEYAG LKVARRQMQD AAGRRHFHAS QCPRPTSPVS TDSNMSAVVI QKARPAKKQK HQPGHLRREA YADDLPPPPV PPPAIKSPTV QSKAQLEVRP VMVPKLASIE ARTDRSSDRK GGSYKGREAL DGRQVTDLRT NPSDPREAQE QPNDGKGRGT RQPKRDLPPA KTHLGQEDIL PYCRPTFPTS NNPRDPSSSS SMSSRGSGSR QREQANVGRR NMAEMQVLGG FERGDENNEE LEETES |