Learn More
enQuireBio™ Recombinant Mouse TNFR Protein
A cDNA sequence encoding the TNFR was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10885-5ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
21937 | |
Greater than 97.0% as determined by SDS-PAGE. | |
Research Use Only | |
Tnfrsf1a | |
Recombinant Protein | |
E. coli | |
IHPSGVTGLV PSLGDREKRD SLCPQGKYVH SKNNSICCTK CHKGTYLVSD CPSPGRDTVC RECEKGTFTA SQNYLRQCLS CKTCRKEMSQ VEISPCQADK DTVCGCKENQ FQRYLSETHF QCVDCSPCFN GTVTIPCKET QNTVCNCHAG FFLRESECVP CSHCKKNEEC MKLCLPPPLA NVTNPQDSGT A |
TNFR Protein | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by its ability to inhibit the TNF-? mediated cytotoxicity in the L-929 cells is less than 1.0 microg/ml, corresponding to a specific activity of > 1000 IU/mg in the presence of 0.1 ng/mL of rMuTNF-a. | |
Mouse | |
Untagged | |
TNFR protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |