Learn More
enQuireBio™ Recombinant Mouse UCHL1 Protein
A cDNA sequence encoding the UCHL1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13879-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
UCHL1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQLKPME INPEMLNKVL AKLGVAGQWR FADVLGLEEE TLGSVPSPAC ALLLLFPLTA QHENFRKKQI EELKGQEVSP KVYFMKQTIG NSCGTIGLIH AVANNQDKLE FEDGSVLKQF LSETEKLSPE DRAKCFEKNE AIQAAHDSVA QEGQCRVDDK VNFHFILFNN VDGHLYELDG RMPFPVNHGA SSEDSLLQDA AKVCREFTER EQGEVRFSAV ALCKAA | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
UCHL1 protein solution (1 mg/ml) containing Phosphate buffered saline, 10% glycerol and 1mM DTT. |