missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VEGF-120 Protein
Recombinant Protein
Marque: enQuireBio™ QP5215-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001273987.1 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKCDKPR R | |
Greater than 95% as determined by SDS-PAGE. |
VEGF-120 Protein | |
Research Use Only | |
VEGFA | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |