missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Multi Species Exendin 4 Protein

A cDNA sequence encoding the Exendin 4 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10593-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15974348

  • 203.00€ / 100µg
Expédition estimée: 24-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Multi Species Exendin 4 Protein
Research Use Only
Regulates Glucose levels rapidly2. Reduces Insulin resistence 3. Reduces Glucagon4. Reduces HbA1c5. Stimulates beta cell growth which stimulates insulin production.
Multi Species
Untagged
The Exendin-4 peptide was lyophilized after extensive dialyses against 20mM PBS and contains 4% mannitol.
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The sequence determined and was found to be His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions