Learn More
enQuireBio™ Recombinant Multi Species PKACa2- RIa2 Protein
A cDNA sequence encoding the PKACa2- RIa2 was constructed and used to recombinantly synthesize the protein.
131.00€ - 5365.00€
Spécification
Nom Multi Species PKACa2- RIa2 Protein
État réglementaire Research Use Only
Concentration en endotoxines < 1.0 EU per ug protein as determined by the LAL method.
Activité biologique Holoenzyme can be activated by adding the second messenger cAMP (Activation constant about 100nM) releasing two monomeric catalytic subunits.
Type de produit Recombinant Protein
Consulter d'autres spécifications
Products 3
Product Code Brand Quantité Price Quantity & Availability  
Product Code Brand Quantité Price Quantity & Availability  
15945278
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP10823-1UG
1 μg
131.00€
1µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 26-06-2024
pour voir le stock disponible 
15955278
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP10823-3UG
3 μg
203.00€
3µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 26-06-2024
pour voir le stock disponible 
15935278
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP10823-100UG
100 μg
5365.00€
100µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 26-06-2024
Specifications
Multi Species PKACa2- RIa2 Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
PKA holoenzyme type-I alpha is supplied in 50% glycerol.
Research Use Only
Holoenzyme can be activated by adding the second messenger cAMP (Activation constant about 100nM) releasing two monomeric catalytic subunits.
Multi Species
Untagged
Greater than 95.0% as determined by SDS-PAGE.
Videos
SDS
Documents
Documents
Certificates