Learn More
enQuireBio™ Recombinant other Borrelia Afzelii p100 Protein
A cDNA sequence encoding the Borrelia Afzelii p100 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11180-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Borrelia Afzelii p100 Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His |
Greater than 80.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
Borrelia Afzelii p100 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |