Learn More
enQuireBio™ Recombinant other Car b 1.0109 Protein
A cDNA sequence encoding the Car b 1.0109 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11274-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Car b 1.0109 Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Car b 1.0109 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
Greater than 80.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGVFNYEAET PSVIPAARLF KSYVLDGDKL IPKVAPQAIS SVENVGGNGG PGTIKNITFA EGSPFKFVKE RVDEVDNANF KYNYTVIEGD VLGDKLEKVS HELKIVAAPG GGSILKISSK FHAKGDHEVN AEEMKGAKEM AEKLLRAVES YLLAHTAEYN |