missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Der P1 Protein
A cDNA sequence encoding the Der P1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11657-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
Der P1 Protein | |
Research Use Only | |
DERP1 | |
Other | |
His | |
(1 mg/ml) 60mM NaCl, 50mM Tris-HCl pH 8.0 and 1.2M Urea. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSIKTFEEYKKAFNKSYATFEDEEAARKNFLESVKYVQSNGGAINHLSDLSLDEFK NRFLMSAEAFEHLKTQFDLNAETNACSINGNAPAEIDLRQMRTVTPIRMQGGCGSAWAFS GVAATESAYLAYRNQSLDLAEQELVDCASQHGCHGDTIPRGIEYIQHNGVVQESYYRYVA REQSCRRPNAQRFGISNYCQIYPPNVNKIREALAQTHSAIAVIIGIKDLDAFRHYDGRTI IQRDNGYQPNYHAVNIVGYSNAQGVDYWIVRNSWDTNWGDNGYGYFAANIDLMMIEEYPY VVILHHHHHH | |
Protein is >95% pure as determined by 10% SDS-PAGE (coomassie staining). |