Learn More
enQuireBio™ Recombinant other DUT Pyrococcus Fruriosus Protein
A cDNA sequence encoding the DUT Pyrococcus Fruriosus was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP10580-2000IU
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15994308
  • 450.00€ / 2000 unités
Expédition estimée: 09-08-2022
Spécification
DUT Pyrococcus Fruriosus Protein
2000 IU
< 1.0 EU per ug protein as determined by the LAL method.
Other
Untagged
Greater than 97.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
dUTPase is supplied in 20mM Tris-HCl (pH 8.2), 1mM DTT, 0.1mM EDTA, 100mM KCl, 0.1% Nonidet P40, 0.1% Tween 20 and 50% glycerol at a concentration of 10U/microL of the enzyme.