Learn More
enQuireBio™ Recombinant other GCV Protein
A cDNA sequence encoding the GCV was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11945-500mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
GCV Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Greater than 99.0%. |
500 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |