missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other GLB1 Protein

A cDNA sequence encoding the GLB1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11980-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15998988

  • 137.00€ / 10µg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

945006
Protein is >95% pure as determined by SDS-PAGE, by measuring optical density at 280 nm and by method of Bradford et al.
Research Use Only
lacZ
Other
Untagged
Beta-Galactosidase (1 mg/1ml) is formulated in 8M urea, 20mM Tris-HCl pH 8.0, and 10mM beta-mercaptoethanol
GLB1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MTMITDSLAV VLQRRDWENP GVTQLNRLAA HPPFASWRNS EEARTDRPSQ QLRSLNGEWR FAWFPAPEAV PESWLECDLP EADTVVVPSN WQMHGYDAPI YTNVTYPITV NPPFVPTENP TGCYSLTFNV DESWLQEGQT RIIFDGVNSA FHLWCNGRWV GYGQDSRLPS EFDLSAFLRA GENRLAVMVL RWSDGSYLED QDMWRMSGIF RDVSLLHKPT TQISDFHVAT RFNDDFSRAV LEAEVQMCGE LRDYLRVTVS LWQGETQVAS GTAPFGGEII DERGGYADRV TLRLNVENPK LWSAEIPNLY RAVVELHTAD GTLIEAEACD VGFREVRIEN GLLLLNGKPL LIRGVNRHEH HPLHGQVMDE QTMVQDILLM KQNNFNAVRC SHYPNHPLWY TLCDRYGLYV VDEANIETHG MVPMNRLTDD PRWLPAMSER VTRMVQRDRN HPSVIIWSLG NESGHGANHD ALYRWIKSVD PSRPVQYEGG GADTTATDII CPMYARVDED QPFPAVPKWS IKKWLSLPGE TRPLILCEYA HAMGNSLGGF AKYWQAFRQY PRLQGGFVWD WVDQSLIKYD ENGNPWSAYG GDFGDTPNDR QFCMNGLVFA DRTPHPALTE AKHQQQFFQF RLSGQTIEVT SEYLFRHSDN ELLHWMVALD GKPLASGEVP LDVAPQGKQL IELPELPQPE SAGQLWLTVR VVQPNATAWS EAGHISAWQQ WRLAENLSVT LPAASHAIPH LTTSEMDFCI ELGNKRWQFN RQSGFLSQMW IGDKKQLLTP LRDQFTRAPL DNDIGVSEAT RIDPNAWVER WKAAGHYQAE AALLQCTADT LADAVLITTA HAWQHQGKTL FISRKTYRID GSGQMAITVD VEVASDTPHP ARIGLNCQLA QVAERVNWLG LGPQENYPDR LTAACFDRWD LPLSDMYTPY VFPSENGLRC GTRELNYGPH QWRGDFQFNI SRYSQQQLME TSHRHLLHAE EGTWLNIDGF HMGIGGDDSW SPSVSAEFQL SAGRYHYQLV WCQK
Vidéos
FDS
Documents
Certificats
Promotions

Promotions