Learn More
enQuireBio™ Recombinant other HSA Protein
A cDNA sequence encoding the HSA was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12314-HIS-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
213 | |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
ALB | |
Other | |
His | |
HSA protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |
HSA Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
insect | |
RGVFRRDAHK SEVAHRFKDL GEENFKALVL IAFAQYLQQC PFEDHVKLVN EVTEFAKTCV ADESAENCDK SLHTLFGDKL CTVATLRETY GEMADCCAKQ EPERNECFLQ HKDDNPNLPR LVRPEVDVMC TAFHDNEETF LKKYLYEIAR RHPYFYAPEL LFFAKRYKAA FTECCQAADK AACLLPKLDE LRDEGKASSA KQRLKCASLQ KFGERAFKAW AVARLSQRFP KAEFAEVSKL VTDLTKVHTE CCHGDLLECA DDRADLAKYI CENQDSISSK LKECCEKPLL EKSHCIAEVE NDEMPADLPS LAADFVESKD VCKNYAEAKD VFLGMFLYEY ARRHPDYSVV LLLRLAKTYE TTLEKCCAAA DPHECYAKVF DEFKPLVEEP QNLIKQNCEL FEQLGEYKFQ NALLVRYTKK VPQVSTPTLV EVSRNLGKVG SKCCKHPEAK RMPCAEDYLS VVLNQLCVLH EKTPVSDRVT KCCTESLVNR RPCFSALEVD ETYVPKEFNA ETFTFHADIC TLSEKERQIK KQTALVELVK HKPKATKEQL KAVMDDFAAF VEKCCKADDK ETCFAEEGKK LVAASQAALG LHHHHHH |