Learn More
enQuireBio™ Recombinant other HYAL1 Protein
A cDNA sequence encoding the HYAL1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12361-1500IU
Informations supplémentaires : Poids : 0.01000kg
Spécification
HYAL1 Protein | |
Research Use Only | |
Recombinant Protein | |
Bovine | |
The enzyme was lyophilized 1xPBS and 2% sucrose. |
1500 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |