Learn More
enQuireBio™ Recombinant other Luciferase Firefly Protein
A cDNA sequence encoding the Luciferase Firefly was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12597-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Luciferase Firefly Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
The Luciferase protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8), 1mM DTT, and 10% glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MMEDAKNIKK GPAPFYPLED GTAGEQLHKA MKRYALVPGT IAFTDAHIEV DITYAEYFEM SVRLAEAMKR YGLNTNHRIV VCSENSLQFF MPVLGALFIG VAVAPANDIY NERELLNSMG ISQPTVVFVS KKGLQKILNV QKKLPIIQKI IIMDSKTDYQ GFQSMYTFVT SHLPPGFNEY DFVPESFDRD KTIALIMNSS GSTGLPKGVA LPHRTACVRF SHARDPIFGN QIIPDTAILS VVPFHHGFGM FTTLGYLICG FRVVLMYRFE EELFLRSLQD YKIQSALLVP TLFSFFAKST LIDKYDLSNL HEIASGGAPL SKEVGEAVAK RFHLPGIRQG YGLTETTSAI LITPEGDDKP GAVGKVVPFF EAKVVDLDTG KTLGVNQRGE LCVRGPMIMS GYVNNPEATN ALIDKDGWLH SGDIAYWDED EHFFIVDRLK SLIKYKGYQV APAELESILL QHPNIFDAGV AGLPDDDAGE LPAAVVVLEH GKTMTEKEIV DYVASQVTTA KKLRGGVVFV DEVPKGLTGK LDARKIREIL IKAKKGGKIA V |