Learn More
enQuireBio™ Recombinant other Myelin Basic Protein
A cDNA sequence encoding the Myelin Basic Protein was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12658-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
7008 | |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
TEF | |
Other | |
His | |
The MBP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |
Myelin Basic Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGNHAGK RELNAEKAST NSETNRGESE KKRNLGELSR TTSEDNEVFG EADANQNNGT SSQDTAVTDS KRTADPKNAW QDAHPADPGS RPHLIRLFSR DAPGREDNTF KDRPSESDEL QTIQEDSAAT SESLDVMASQ KRPSQRHGSK YLATASTMDH ARHGFLPRHR DTGILDSIGR FFGGDRGAPK RGSGKVSSEE |