Learn More
enQuireBio™ Recombinant other Protease Protein
A cDNA sequence encoding the Protease was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13159-1,000IU
Informations supplémentaires : Poids : 0.01000kg
Spécification
Protease Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli |
1000 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |