Learn More
enQuireBio™ Recombinant other protein 1
A cDNA sequence encoding the Protein A was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10853-EC-10mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
protein 1 | |
10 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
7 mg human IgG/mg protein-A. | |
Other | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution containing no additives. |
Greater than 98.0% as determined by SDS-PAGE & RP-HPLC. | |
Research Use Only | |
spa | |
Recombinant Protein | |
E. coli | |
MKKKNIYSIR KLGVGIASVT LGTLLISGGV TPAANAAQHD EAQQNAFYQV LNMPNLNADQ RNGFIQSLKD DPSQSANVLG EAQKLNDSQA PKADAQQNKF NKDQQSAFYE ILNMPNLNEE QRNGFIQSLK DDPSQSTNVL GEAKKLNESQ APKADNNFNK EQQNAFYEIL NMPNLNEEQR NGFIQSLKDD PSQSANLLAE AKKLNESQAP KADNKFNKEQ QNAFYEILHL PNLNEEQRNG FIQSLKDDPS QSANLLAEAK KLNDAQAPKA DNKFNKEQQN AFYEILHLPN LTEEQRNGFI QSLKDDPSVS KEILAEAKKL NDAQAPKEED NNKPGKEDGN KPGKEDGNKP GKEDNKKPGK EDGNKPGKED NKKPGKEDGN KPGKEDGNKP GKEDGNKPGK EDGNKPGKED GNGVHVVKPG DTVNDIAKAN GTTADKIAAD NKLADKNMIK PGQELVVDKK QPANHADANK AQALPETGEE NPFIGTTVFG GLSLALGAAL LAGRRREL |