missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Thymosin ?1 Protein
A cDNA sequence encoding the Thymosin ?1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13745-5mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
Thymosin ?1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSDAAVDTSS EITTKDLKEK KEVVEEAENG RDAPANGNAE NEENGEQEAD NEVDEEEEEG GEEEEEEEEG DGEEEDGDED EEAESATGKR AAEDDEDDDV DTKKQKTDED D | |
Greater than 99.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
5 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The protein (1 mg/ml) was lyophilized with no additives. |