Learn More
enQuireBio™ Recombinant other Urease Protein
A cDNA sequence encoding the Urease was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP10907-50mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15995528
  • 1425.00€ / 50mg
Expédition estimée: 10-05-2024
Spécification
Urease Protein
Research Use Only
The activity was found to be 141U/mg powder.
Other
Untagged
Greater than 95.0% as determined by SDS-PAGE.
50 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
Each mg of protein contains 420microg Potassium Phosphate and 30microg EDTA Na2.