Learn More
enQuireBio™ Recombinant Pig Calpain-2 catalytic subunit Protein
A DNA sequence encoding the Sus scrofa (Pig) Calpain-2 catalytic subunit, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5757-ec-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Calpain-2 catalytic subunit Protein | |
100 μg | |
Not determined. | |
Full Length | |
Pig | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CAPN2 | |
Recombinant Protein | |
E. coli | |
YPNTFWMNPQ YLIKLEEEDE DQEDGESGCT FLVGLIQKHR RRQRKMGEDM HTIGFGIYEV PEELTGQTNI HLSKNFFLTH RARERSDTFI NLREVLNRFK LPPGEYILVP STFEPNKDGD FCIRVFSEKK ADYQVVDDEI EADLEENDAS EDDIDDGFRR LFAQLAGEDA EISAFELQTI LRRVLAKRQD IKSDGFSIET CRIMVDMLDS DGSAKLGLKE FYILWTKIQK YQKIYREIDV DRSGTMNSYE MRKALEEAGF KLPCQLHQVI VARFADDQLI IDFDNFVRCL VRLETLFRIS KQLDSENTGT IELDLISWLC FSVL |