missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Adipsin / Complement Factor D / CFD Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Adipsin / Complement Factor D / CFD, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP5823-ye-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15988019

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet


Description

Description

Spécification

Spécification

NP_001071110.1
Adipsin / Complement Factor D / CFD Protein
Research Use Only
Cfd
Recombinant Protein
Yeast
ILGGQEAMAH ARPYMASVQV NGTHVCGGTL VDEQWVLSAA HCMDGVTKDE VVQVLLGAHS LSSPEPYKHL YDVQSVVLHP GSRPDSVEDD LMLFKLSHNA SLGPHVRPLP LQREDREVKP GTLCDVAGWG VVTHAGRRPD VLQQLTVSIM DRNTCNLRTY HDGAITKNMM CAESNRRDTC RGDSGGPLVC GDAVEAVVTW GSRVCGNRRK PGVFTRVATY VPWIENVLSG NVSVNVTA
Greater than 82.10% as determined by SDS-PAGE.
54249
10 μg
Not determined.
Full Length of Mature Protein
Rat
His
Tris-based buffer, 50% glycerol
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions