Learn More
enQuireBio™ Recombinant Rat Alkaline Phosphatase / ALPL Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Alkaline Phosphatase / ALPL, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5647-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_037191.1 | |
Alkaline Phosphatase / ALPL Protein | |
1 mg | |
Not determined. | |
Full Length of Mature Protein | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
25586 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Alpl | |
Recombinant Protein | |
E. coli | |
FVPEKEKDPS YWRQQAQETL KNALKLQKLN TNVAKNIIMF LGDGMGVSTV TAARILKGQL HHNTGEETRL EMDKFPFVAL SKTYNTNAQV PDSAGTATAY LCGVKANEGT VGVSAATERT RCNTTQGNEV TSILRWAKDA GKSVGIVTTT RVNHATPSAA YAHSADRDWY SDNEMPPEAL SQGCKDIAYQ LMHNIKDIDV IMGGGRKYMY PKNRTDVEYE LDEKARGTRL DGLDLISIWK SFKPRHKHSH YVWNRTELLA LDPSRVDYLL GLFEPGDMQY ELNRNNLTDP SLSEMVEVAL RILTKNPKGF FLLVEGGRID HGHHEGKAKQ ALHEAVEMDE AIGKAGTMTS QKDTLTVVTA DHSHVFTFGG YTPRGNSIFG LAPMVSDTDK KPFTAILYGN GPGYKVVDGE RENVSMVDYA HNNYQAQSAV PLRHETHGGE DVAVFAKGPM AHLLHGVHEQ NYIPHVMAYA SCIGANLDHC AWAS |