missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Allograft inflammatory factor 1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Allograft inflammatory factor 1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5632-ye-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_058892.1 | |
Allograft inflammatory factor 1 Protein | |
Research Use Only | |
Aif1 | |
Recombinant Protein | |
Yeast | |
SQSKDLQGGK AFGLLKAQQE ERLDGINKHF LDDPKYSSDE DLQSKLEAFK TKYMEFDLNG NGDIDIMSLK RMLEKLGVPK THLELKKLIR EVSSGSEETF SYSDFLRMML GKRSAILRMI LMYEEKNKEH QKPTGPPAKK AISELP | |
Greater than 90% as determined by SDS-PAGE. |
29427 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |