Learn More
enQuireBio™ Recombinant Rat Allograft inflammatory factor 1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Allograft inflammatory factor 1, was expressed in the hosts and tags indicated.
Marque:  enQuireBio™ QP5632-ye-1mg
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15996579
  • 3260.00€ / 1mg
Expédition estimée: 28-06-2024
Spécification
NP_058892.1
Allograft inflammatory factor 1 Protein
Research Use Only
Aif1
Recombinant Protein
Yeast
SQSKDLQGGK AFGLLKAQQE ERLDGINKHF LDDPKYSSDE DLQSKLEAFK TKYMEFDLNG NGDIDIMSLK RMLEKLGVPK THLELKKLIR EVSSGSEETF SYSDFLRMML GKRSAILRMI LMYEEKNKEH QKPTGPPAKK AISELP
Greater than 90% as determined by SDS-PAGE.
29427
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol