Learn More
enQuireBio™ Recombinant Rat bFGF / FGF2 Protein
Recombinant Protein
Marque:  enQuireBio™ QP5493-50ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15926249
  • 259.00€ / 50µg
Expédition estimée: 27-06-2022
Spécification
NP_062178.1
bFGF / FGF2 Protein
50 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 1 ng/mL in the BALB/c 3T3 proliferation assay. The bioactivity acceptance criteria is set at 1.0 x 10^6 units/mg.
Rat
Untagged
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
54250
Greater than 95% as determined by SDS-PAGE.
Research Use Only
FGF2
Recombinant Protein
E. coli
MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS