missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat bFGF / FGF2 Protein
Recombinant Protein
Marque: enQuireBio™ QP5493-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_062178.1 | |
bFGF / FGF2 Protein | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 1 ng/mL in the BALB/c 3T3 proliferation assay. The bioactivity acceptance criteria is set at 1.0 x 10^6 units/mg. | |
Rat | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |
54250 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
FGF2 | |
Recombinant Protein | |
E. coli | |
MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS |