missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IGF1 / IGF-I Protein
Recombinant Protein
Marque: enQuireBio™ QP5492-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001075946.2 | |
IGF1 / IGF-I Protein | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IGF1 / IGF-I induces MCF-7 cell proliferation (typical ED50 is ? 100 ng/ml). | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
24482 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
IGF1 | |
Recombinant Protein | |
E. coli | |
GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA |