Learn More
enQuireBio™ Recombinant Rat IL 17E Protein
A cDNA sequence encoding the IL 17E was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12393-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
IL 17E Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
Untagged | |
Lyophilized from a concentrated (1 mg/ml) solution containing no additives. |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
CSHLPRCCPS KQQEFPEEWL KWNPAPVSPP EPLRHTHHPE SCRASKDGPL NSRAISPWSY ELDRDLNRVP QDLYHARCLC PHCVSLQTGS HMDPMGNSVP LYHNQTVFYR RPCHGEQGAH GRYCLERRLY RVSLACVCVR PRMMA |