missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Plasma kallikrein Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Plasma kallikrein, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP6269-ye-500ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16032080

  • 2175.00€ / 500µg
Expédition estimée: 02-11-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Plasma kallikrein Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Klkb1
Recombinant Protein
Yeast
IVGGTNSSLG EWPWQVSLQV KLVSQNHMCG GSIIGRQWIL TAAHCFDGIP YPDVWRIYGG ILNLSEITNK TPFSSIKELI IHQKYKMSEG SYDIALIKLQ TPLNYTEFQK PICLPSKADT NTIYTNCWVT GWGYTKERGE TQNILQKATI PLVPNEECQK KYRDYVITKQ MICAGYKEGG IDACKGDSGG PLVCKHSGRW QLVGITSWGE GCARKEQPGV YTKVAEYIDW ILEKIQSSKE RALETSPA
Vidéos
FDS
Documents
Certificats
Promotions

Promotions