Learn More
enQuireBio™ Recombinant Rat S100G Protein
A cDNA sequence encoding the S100G was constructed and used to recombinantly synthesize the protein.
131.00€ - 3190.00€
Specifications
Identification génétique (Entrez) 24249
Nom S100G Protein
État réglementaire Research Use Only
Concentration en endotoxines Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Symbole de gène(s) S100g
View More Specs
Products 3
Product Code Brand Quantité Price Quantity & Availability  
Product Code Brand Quantité Price Quantity & Availability  
15943469
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP13406-1UG
1 μg
131.00€
1µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 21-06-2024
pour voir le stock disponible 
15953469
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP13406-5UG
5 μg
203.00€
5µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 21-06-2024
pour voir le stock disponible 
15933469
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP13406-100UG
100 μg
3190.00€
100µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 21-06-2024
Specifications
24249
Research Use Only
S100g
Rat
His
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 100mM Phosphate buffer, pH 7.3.
S100G Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSAKKSPEEM KSIFQKYAAK EGDPNQLSKE ELKLLIQSEF PSLLKASSTL DNLFKELDKN GDGEVSYEEF EVFFKKLSQ
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents
Documents
Certificates