missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rhesus Macaque CCL17/TARC Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
1 mg
10 μg
100 μg
500 μg
Conditionnement:
100µg
10µg
1mg
500µg
Description
Fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 1.0-10 ng/mL.
Spécification
Spécification
| À utiliser avec (application) | Bioactivity |
| Formule | Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, 300 mM NaCl, pH 7.4, with 0.02 % Tween-20. |
| Identification génétique (Entrez) | 6361 |
| Poids moléculaire | MolecularWeight-theroretical: 8.1 kDa |
| Quantité | 500 μg |
| Conditions de stockage | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Concentration en endotoxines | Less than 0.1 EU/μg of CCL17/TARC as determined by LAL method. |
| Alias de gène | ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine |
| Symbole de gène(s) | CCL17 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu