Learn More
Thermo Scientific™ Human RET [V804L], GST Tag Recombinant Protein
The receptor tyrosine kinase RET is a member of the cadherin superfamily. RET plays a crucial role in neural crest development, and can undergo oncogenic activation by cytogenetic rearrangement.
Marque: Thermo Scientific™ PV4433
Code nomenclature Nacres: NA.24
Informations supplémentaires : Poids : 0.00900kg
Description
- The receptor tyrosine kinase RET is a member of the cadherin superfamily
- RET plays a crucial role in neural crest development, and can undergo oncogenic activation by cytogenetic rearrangement
- The V804L mutant has been observed in association with cancer
Spécification
NP_066124, V804L | |
79 kDa | |
RET V804L | |
Human | |
GST Tag | |
Tyrosine Kinases | |
Baculovirus | |
Frozen |
Kinase Assay | |
1 mg | |
RET Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Cytoplasmic | |
Drug Development |