Learn More
Microbiologics™ Salmonella enterica subsp. enterica serovar Abony derived from NCTC 6017
Microbiologics Salmonella enterica subsp. enterica serovar Abony derived from NCTC 6017 has a simple, all-inclusive design that makes life easier for lab techs while reducing chances for errors
Marque: Microbiologics™ 0890K
Code nomenclature Nacres: NA.81
Informations supplémentaires : Poids : 0.50000kg Transport : UN number : 3373 Chem class : 6.2
Description
- Biosafety Level: 2
- 6 self-contained units of a single organism
- Serovar Abony
- Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab
Spécification
Salmonella enterica subsp. enterica serovar abony | |
NCTC 6017 | |
KWIK-STIK | |
Microbial Identification | |
6 |
2 | |
2°C to 8°C | |
United States | |
Plastic Canister |