missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV-2 ORF8 (strain Wuhan-Hu-1) Polyclonal Antibody
GREENER_CHOICE

Code produit 17890766
Change view
Click to view available options
Quantité:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
17890766 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17890766 Supplier Invitrogen™ Supplier No. PA5143463

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The structure of the SARS-CoV-2 ORF8 protein involves two novel intermolecular interfaces layered onto an ORF7 fold. One is mediated by a disulfide bond, the other is noncovalent, and both are novel approximately 60-residue core is similar to SARS-CoV-2 ORF7a. The addition of two dimerization interfaces is unique to SARS-CoV-2 ORF8.
TRUSTED_SUSTAINABILITY

Specifications

Antigène SARS-CoV-2 ORF8 (strain Wuhan-Hu-1)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugué Unconjugated
Formule PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Expression ORF8
Alias de gène COVID19 ORF8
Symboles de gène(s) ORF8
Espèces hôtes Rabbit
Immunogène Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of SARS-CoV-2 (COVID-19) ORF8 (Strain:Wuhan-Hu-1). The exact sequence is proprietary.
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 43740577
Espèces cibles Virus
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.