Learn More
ACROBiosystems SARS-CoV-2 S protein RBD (K417T, E484K, N501Y), Fc Tag
Marque:  ACROBiosystems SPD-C5258-1mg
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 17256510
  • 3261.73€ / 1mg
Expédition estimée: 17-10-2022
pour voir le stock disponible 
Description
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.1 kDa.
Spécification
Spécification
Powder
1 mg
Arg 319 - Lys 537
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
Unconjugated
HEK293
SARS-CoV-2
51.1 kDa
SARS-CoV-2 S protein RBD (K417T,E484K,N501Y), Fc Tag (SPD-C5258) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (K417T, E484K, N501Y)).
-20°C
S protein RBD
Fc Tag
Viral Proteins