Learn More
ACROBiosystems SARS-CoV-2 S protein RBD (K417T), His Tag
Marque:  ACROBiosystems SPD-C52Ht-100ug
Informations supplémentaires : Poids : 0.00010kg
Code produit. 17246530
  • 483.87€ / 100µg
Expédition estimée: 24-05-2024
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa.
Spécification
Spécification
QHD43416.1 (K417T)
100 μg
Arg 319 - Lys 537
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
Unconjugated
His Tag
Powder
26.5 kDa
SARS-CoV-2 S protein RBD (K417T), His Tag (SPD-C52Ht) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (K417T)).
-20°C
S Protein RBD
SARS-CoV-2
HEK293
Viral Proteins