Learn More
ACROBiosystems SARS-CoV-2 S protein RBD (K417T), His Tag
Marque:  ACROBiosystems SPD-C52Ht-100ug
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 17246530
  • 479.87€ / 100µg
Expédition estimée: 11-04-2024
pour voir le stock disponible 
Description
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa.
Spécification
Spécification
QHD43416.1 (K417T)
100 μg
Arg 319 - Lys 537
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
Unconjugated
His Tag
Powder
26.5 kDa
SARS-CoV-2 S protein RBD (K417T), His Tag (SPD-C52Ht) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (K417T)).
-20°C
S Protein RBD
SARS-CoV-2
HEK293
Viral Proteins