Learn More
ACROBiosystems SARS-CoV-2 S1 protein NTD (HV69-70del, Y144del), His Tag
Marque:  ACROBiosystems S1D-C52Hd-1mg
Informations supplémentaires : Poids : 0.00010kg
Code produit. 17256450
  • 2905.82€ / 1mg
Expédition estimée: 21-07-2022
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.5 kDa.
Spécification
Spécification
QHD43416.1 (HV69-70 del, Y144del)
34.5 kDa
SARS-CoV-2 S1 protein NTD (HV69-70 del, Y144del), His Tag (S1D-C52Hd) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1 (HV69-70 del, Y144del)).
-20°C
S1 protein NTD
His Tag
Viral Proteins
Powder
1 mg
Ser 13 - Leu 303
S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD,COVID-19
Unconjugated
HEK293
SARS-CoV-2