Learn More
ACROBiosystems SARS-CoV-2 S1 protein NTD (HV69-70del, Y144del), His Tag
Marque:  ACROBiosystems S1D-C52Hd-100ug
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 17246450
  • 456.19€ / 100µg
Expédition estimée: 10-01-2023
pour voir le stock disponible 
Description
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.5 kDa.
Spécification
Spécification
QHD43416.1 (HV69-70 del, Y144del)
34.5 kDa
SARS-CoV-2 S1 protein NTD (HV69-70 del, Y144del), His Tag (S1D-C52Hd) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1 (HV69-70 del, Y144del)).
-20°C
S1 protein NTD
His Tag
Viral Proteins
Powder
100 μg
Ser 13 - Leu 303
S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD,COVID-19
Unconjugated
HEK293
SARS-CoV-2