Learn More
Abnova™ SERPINA3 (Human) Recombinant Protein
Human SERPINA3 full-length ORF ( AAH03559, 25 a.a. - 423 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00000012-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: PNSPLDEENLTQENQDRGTHVDLGLASANVDFAFSLYKQLVLKAPDKNVIFSPLSISTALAFLSLGAHNTTLTEILKGLKFNLTETSEAEIHQSFQHLLRTLNQSSDELQLSMGNAMFVKEQLSLLDRFTEDAKRLYGSEAFATDFQDSAAAKKLINDYVKNGTRGKITDLIKDLDSQTMMVLVNYIFFKAKWEMPFDPQDTHQSRFYLSKKKWVMVPMMSLHHLTIPYFRDEELSCTVVELKYTGNASALFILPDQDKMEEVEAMLLPETLKRWRDSLEFREIGELYLPKFSISRDYNLNDILLQLGIEEAFTSKADLSGITGARNLAVSQVVHKAVLDVFEEGTEASAATAVKITLLSALVETRTIVRFNRPFLMIIVPTDTQNIFFMSKVTNPKQA
Spécification
AAH03559 | |
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SERPINA3 | |
GST |
12 | |
Wheat germ expression system | |
10 μg | |
AACT, ACT, GIG24, GIG25, MGC88254 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |