missing translation for 'onlineSavingsMsg'
Learn More

SHIELD Scientific SHIELDskin™ ORANGE NITRILE™ 260

Encompass_Preferred
Click to view available options
Dimensions:
X-Small
Small
Medium
Large
X-Large
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

SHIELDskin™ gloves provide you with a high level of compliance, comfort, and protection. These nitrile/neoprene gloves are specifically designed to protect assays from contamination and protect you from biological and chemical hazards using the revolutionary twinSHIELD™ technology .

  • Brand signature: Orange for biological risk
  • Laboratory gloves for biohazard protection
  • Latex gloves and nitrile/neoprene gloves
  • Non-sterile and sterile gloves
  • Maximum comfort
  • Compliance with the latest European standards
  • Category III PPE gloves (PPE Regulation (EU) 2016/425)
  • Class 1 MDR (Regulation (EU) 2017/745)*
  • Extra length from 260 to 300 mm (meets or exceeds ISO 21420:2020+A1:2022)
  • Biohazard: ISO 374-5:2016
  • Biosafety: AQL 0.25 (ISO 374-2:2019 Level 3)
  • Virus resistant (ISO 16604:2004 Procedure B)
  • Extensively tested for chemical permeation according to ISO 374-1:2016+A1:2018 (Test method EN 16523-1:2015+A1:2018)
  • Reduced risk of allergies
  • Double-walled protection afforded by the twinSHIELD™ technology
  • Packaging designed to comply with laboratory environments processes
Certifications/Conformité AQL 0.25
Certifications / conformité (Europe) PPER (EU) 2016/425, ISO 21420:2020, EN455-2:2015, ISO 374-1:2016-A1:2018
Accelerator Free Yes
Applications Laboratory
AQL 0.25
Couleur Orange
Jetable Single-use
Finition externe Fingertip Textured
Finger Thickness (English) 6.7 mil
Finger Thickness (Metric) 0.17 mm
Latex Free Yes
Matériau Nitrile, Neoprene
Palm Thickness (English) 5.5 mil
Palm Thickness (Metric) 0.14 mm
Poudré Powder Free
Type de produit Protective Gloves
Dimensions Medium
Stérilité Non-sterile
Épaisseur (impériale) 3.9 mil
Épaisseur (métrique) 0.10 mm
Longueur (impériale) 10.2 in.
Longueur (métrique) 260 mm
Afficher plus Afficher moins
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'