Learn More
Invitrogen™ Sifalimumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Code produit 30284589
Change view
Click to view available options
Quantité:
1 mg
100 μg
Conditionnement:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30284589 100 μg 100µg
30284612 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30284589 Fournisseur Invitrogen™ Code fournisseur MA558635
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sifalimumab is a fully human monoclonal antibody targeting interferon-alpha. The levels of interferon-alpha are elevated in many patients with active systemic lupus erythematosus (SLE, or lupus) and other autoimmune disorders, and may be associated with disease activity. Sifalimumab may suppress the abnormal immune activity associated with lupus by binding to multiple interferon-alpha subtypes seen in the serum of lupus patients.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Sifalimumab
Applications ELISA, Flow Cytometry, Functional Assay
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Alias de gène MDX-1103; MEDI-545
Espèces hôtes Human
Méthode de purification Protein A
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Human
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Lyophilized
Isotype IgG1