Learn More
Oxyde d’argent(II), 98 %, Thermo Scientific Chemicals
Code produit. p-7049783
Click to view available options
Quantité:
5 g
25 g
100 g
Conditionnement:
100g
25g
5g
Code produit. 11338087
Marque: Thermo Scientific Alfa Aesar 022695.06
Demander un format en vrac ou personnalisé
Fabrication de piles alcalines à base d’oxyde d’argent et de zinc
Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 1301-96-8
Formule moléculaire AgO
Poids moléculaire (g/mol) 123.867
Numéro MDL MFCD00044285
Clé InChI OTCVAHKKMMUFAY-UHFFFAOYSA-N
Synonyme silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag
CID PubChem 92152
Nom IUPAC oxoargent
SMILES O=[Ag]
Spécification
Nom chimique ou matériau Silver(II) oxide
Point de fusion >100°C (decomposition)
Couleur Noir à gris
Forme physique Poudre
Quantité 5 g
Numéro UN UN3085
Merck Index 14,8522
Informations sur la solubilité Insoluble in water; Soluble in alkalies,ammonium hydroxide with decomposition and N2 formation.
Poids de la formule 123.87
Pourcentage de pureté 98%
Odeur Odorless
Sensibilité Light and moisture sensitive
Densité 7.44 g/mL