missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNRPN Polyclonal Antibody

Code produit 13424557
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
13424557 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 13424557 Fournisseur Invitrogen™ Code fournisseur PA530225

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, mouse heart. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (97%), Pig (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is one polypeptide of a small nuclear ribonucleoprotein complex and belongs to the snRNP SMB/SMN family. The protein plays a role in pre-mRNA processing, possibly tissue-specific alternative splicing events. Although individual snRNPs are believed to recognize specific nucleic acid sequences through RNA-RNA base pairing, the specific role of this family member is unknown. The protein arises from a bicistronic transcript that also encodes a protein identified as the SNRPN upstream reading frame (SNURF). Multiple transcription initiation sites have been identified and extensive alternative splicing occurs in the 5' untranslated region. Additional splice variants have been described but sequences for the complete transcripts have not been determined. The 5' UTR of this gene has been identified as an imprinting center. Alternative splicing or deletion caused by a translocation event in this paternally-expressed region is responsible for Angelman syndrome or Prader-Willi syndrome due to parental imprint switch failure.
TRUSTED_SUSTAINABILITY

Spécification

Antigène SNRPN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Expression SNRPN
Numéro d’ordre du gène P63162, P63163, P63164
Alias de gène 2410045I01Rik; DKFZp686C0927; DKFZp686M12165; DKFZp761I1912; DKFZp762N022; FE 294; FE 294 psi; FE294; FE294 gene for snRNP-associated polypeptide N; FE294 psi; FE294 psi pseudogene; FLJ33569; FLJ36996; FLJ39265; HCERN3; MGC29886; Peg4; PWCR; Pwcr1; RT-LI; Sm protein D; sm protein N; small nuclear ribonucleoparticle-associated protein (snRNP) mRNA clone Sm51; small nuclear ribonucleoparticle-associated protein (snRNP) mRNA, clone Sm51; small nuclear ribonucleoprotein N; small nuclear ribonucleoprotein polypeptide N; small nuclear ribonucleoprotein-associated protein N; Sm-D; Smn; sm-N; snRNP-N; Snrpn; SNRPN upstream reading frame; SNURF; SNURF-SNRPN; tissue-specific splicing protein; tissue-specific-splicing protein
Symboles de gène(s) SNRPN
Espèces hôtes Rabbit
Immunogène Recombinant protein encompassing a sequence within the center region of human SNRPN. The exact sequence is proprietary.
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 20646, 6638, 81781
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.