missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiosulfate de sodium, 0,1 M, Chem-Lab
Click to view available options
Quantité:
500 mL
1 L
10 L
Description
The thiosulfate anion reacts stoichiometrically with iodine in aqueous solution. Due to the quantitative nature of this reaction, sodium thiosulfate is used as a redox titrant in volumetric analysis and is a component of iodine clock experiments. It can be used to measure the oxygen content of water and to estimate the concentration of certain compounds in solution (hydrogen peroxide, chlorine). It also has many medicinal uses.
Spécification
Point d’ébullition | 100°C |
Nom chimique ou matériau | Sodium Thiosulfate Pentahydrate |
Point de fusion | 48.3°C |
Forme physique | Liquid |
Quantité | 500 mL |
Informations sur la solubilité | Soluble in water |
Poids de la formule | 248.18g/mol |
Concentration or Composition (by Analyte or Components) | 0.1mol/L |
Densité | 1.00g/mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'