Learn More
Invitrogen™ Spartalizumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Code produit 30284540
Change view
Click to view available options
Quantité:
1 mg
100 μg
Conditionnement:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30284540 100 μg 100µg
30282429 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30284540 Fournisseur Invitrogen™ Code fournisseur MA558363
Please to purchase this item. Need a web account? Register with us today!
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Spartalizumab (PDR001) is a humanized IgG4 monoclonal antibody that targets PD-1 with sub-nanomolar affinity to block its interaction with PD-L1/PD-L2, thereby preventing PD-1 mediated inhibitory signals Transduction leads to the activation of T cells.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Spartalizumab Humanized
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Alias de gène NPVPDR001; NVS240118; PDR001
Espèces hôtes Human
Méthode de purification Protein A
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Human
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Lyophilized
Isotype IgG4SP